ContactUsLinkComponent
VWR* Thermometer Armor Case
VWR® Thermometer Armor Cases
Catalog #: 61222-702
Supplier:  VWR International
VWR* Thermometer Armor Case
VWR® Thermometer Armor Cases
Catalog #: 61222-702
Supplier:  VWR International
Specifications
  • Cat. no.:
    61222-702
  • For Use With:
    305 mm (12") Thermometer
Specifications
About this item
These nickel-plated brass armor cases protect thermometers from accidental breakage during use and storage.
Lower section surrounding bulb has small openings for circulation
Available in five lengths to fit SAMA general-purpose total and partial immersion thermometers, including fluoropolymer resin-coated thermometers.
Fitted with screw cap and ring.
Customers Who Bought This Also Bought