Specifications
- Description:Refrigerator/Freezer Thermometer
- Temperature accuracy:±1°C
- Temperature resolution:1°C
- Cat. no.:61161-364
- Probe:Bottle probe (glycol)
- Range:–50…+70 °C (–58…+158 °F)
Specifications
About this item
Accurately monitor temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with this enclosed temperature-buffered sensor.
- Probe is sealed in glycol solution
- Min/max memory
- High and low settable alarms
Bottle insulates sensor from rapid temperature changes when refrigerator door is opened.
Triple display simultaneously shows high, low, and current temperatures. Min/max monitors high/low readings overnight, on weekends, or for any time period—a significant advantage over current reading-only glass thermometers. Range is -58 to 158°F and -50 to 70°C with a resolution of 1° and accuracy of ±1°C.
Alarm feature provides alert when temperature rises above or falls below a set point. Alarm is programmable in 1° increments. Visual and audible signals continue even if temperature returns to non-alarm range. Ten-foot micro-cable permits refrigerator doors to close on it.
Solid-state probe eliminates mercury contamination in refrigerators. Probe is sealed in a miniature bottle (1 x 2-1/2 inches) filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration). Eliminates concerns about contact with food or drinking water.