SP Bel-Art Serum Vial Racks, Polypropylene, Bel-Art Products, a part of SP
Catalog #: 60985-498
Supplier: Bel-Art Products, a Part of SP
Specifications
- Cat. no.:60985-498
- No. of Places:40
- W×D×H:25.4 x 11 x 9 cm (10 x 411/32 x 317/32")
- For Tube O.D.:16 - 20 mm
- Supplier No.:185132040
Specifications
About this item
Just the right size for serum vials, RIA vials and T3/T4 reaction vials.
- Polypropylene rack fabrication
- Corrosion-resistant
- Long effective lifepsan
- Autoclavable
Long-lasting and corrosion-resistant, these polypropylene racks fit most water baths for elevated temperature incubations. Top shelf height is 41 mm (15⁄8"). End plates have handles.
Customers Who Bought This Also Bought