VWR® Plate Paddles
Catalog #: 60941-128
Supplier: VWR International
Specifications
- Description:VWR® plate paddles, plastic
- Cat. no.:60941-128
Specifications
About this item
An easy alternative to rollers, these plate paddles are designed to provide secure sealing around all wells on a microplate.
- Alternative to rollers
- Secure seal around all microplate wells
- Especially effective with raised-rim microplates
Paddles are especially recommended when applying films to raised-rim microplates due to their precise fit within the rim of the plate.
Customers Who Bought This Also Bought