Nalgene® Autoclaving Baskets, Polypropylene, Thermo Scientific
Catalog #: 60915-390
Supplier: Thermo Fisher Scientific
Specifications
- Dimensions:15.2L x 12.4D x 10.5H cm (6 x 47/8 x 41/8")
- Cat. no.:60915-390
- Supplier No.:6917-0127
Specifications
About this item
These baskets hold labware during autoclaving and washing
They allow complete venting and draining. Smooth surface will not scratch plastic or glass. White polypropylene will not rust. Nestable for storage.
Customers Who Bought This Also Bought