Order Entry
ContactUsLinkComponent
NALGENE* Unwire* Polyketone Rack
Nalgene® Unwire™ Racks, ResMer® Resin, Thermo Scientific
Catalog #: 60915-095
NALGENE* Unwire* Polyketone Rack
Nalgene® Unwire™ Racks, ResMer® Resin, Thermo Scientific
Catalog #: 60915-095

Specifications

  • Color:
    Blue
  • Cat. no.:
    60915-095
  • L×W×H:
    28.3 x 11 x 8.3 cm (11 x 41/4 x 31/4")
  • For Tube O.D.:
    30 mm
  • Supplier No.:
    5970-0330

Specifications

About this item

These durable, brightly colored racks feature a three-tier design

Racks for 13mm and 16mm tubes hold 72 tubes; racks for 20mm and 25mm tubes hold 40 tubes; racks for 30mm tubes holds 24 tubes. Autoclavable.

They have excellent chemical resistance and can be immersed in a water bath. Letters and numbers on top tier aid in tube identification. Labels can be placed on large end plates. ResMer® technology allows these racks to be used over a wide temperature range.

Customers Who Bought This Also Bought