Order Entry
ContactUsLinkComponent
NALGENE* Unwire* Polyketone Rack
Nalgene® Unwire™ Racks, ResMer® Resin, Thermo Scientific
Catalog #: 60914-729
NALGENE* Unwire* Polyketone Rack
Nalgene® Unwire™ Racks, ResMer® Resin, Thermo Scientific
Catalog #: 60914-729

Specifications

  • Color:
    Yellow
  • Cat. no.:
    60914-729
  • L×W×H:
    24.8 x 12.7 x 7 cm (9.7 x 5 x 23/4")
  • For Tube O.D.:
    16 mm
  • Supplier No.:
    5970-0216

Specifications

About this item

These durable, brightly colored racks feature a three-tier design

Racks for 13mm and 16mm tubes hold 72 tubes; racks for 20mm and 25mm tubes hold 40 tubes; racks for 30mm tubes holds 24 tubes. Autoclavable.

They have excellent chemical resistance and can be immersed in a water bath. Letters and numbers on top tier aid in tube identification. Labels can be placed on large end plates. ResMer® technology allows these racks to be used over a wide temperature range.

Customers Who Bought This Also Bought