Order Entry
ContactUsLinkComponent
PYREX* Culture Tube
PYREX® Culture Tubes, Reusable, Borosilicate Glass, Corning
Catalog #: 60820-240
Supplier:  Corning
Product available on GSA Advantage®
PYREX* Culture Tube
PYREX® Culture Tubes, Reusable, Borosilicate Glass, Corning
Catalog #: 60820-240
Supplier:  Corning
Product available on GSA Advantage®

Specifications

  • Capacity:
    27 ml
  • Ø:
    18 mm
  • Height:
    150 mm
  • Supplier No.:
    9820-18

Specifications

About this item

Made of PYREX® borosilicate glass, rimless.

Tube 60820-104 can be used for Kahn tests. Tube 60820-126 can be used for Wasserman tests.

  • Very good chemical resistance
  • Very good temperature resistance
  • Ends are fire-polished

Made of PYREX® borosilicate glass, rimless.

Culture tubes have walls and bottom of a uniform thickness. All tubes feature a white marking spot, with the exception of 6 and 22 mm tubes.

Tube 5 ml can be used for Kahn tests. Tube 9 ml can be used for Wasserman tests.

Customers Who Bought This Also Bought