Specifications
- Description:pH Paper, Hydrion MicroFine Disp
- Cat. no.:60775-966
- pH Range:6.0 - 8.0
- Supplier No.:MF-1607
Specifications
About this item
This short-range Hydrion MicroFine pH Paper measures pH solutions in the acid pH range of 2.9-5.2 and offers clear bright single color matches at every 0.3 to 0.4 interval.
- Provides precise, pinpoint pH measurement over a narrow pH range
- Ultra-fast results with accuracy in intervals of 0.2 - 0.3 pH
- Fit easily in to small tubes
These are ideally suited to testing minute amounts of solution, only requiring 0.0001mL for an accurate measurement.