Order Entry
ContactUsLinkComponent
Cylinder Wall Bracket
VWR® Talon® Cylinder Wall Brackets
Catalog #: 60141-124
Supplier:  VWR International
Product available on GSA Advantage®
Cylinder Wall Bracket
VWR® Talon® Cylinder Wall Brackets
Catalog #: 60141-124
Supplier:  VWR International
Product available on GSA Advantage®

Specifications

  • Description:
    Wall Bracket with Strap
  • Height:
    11.8 cm (45/8")
  • Cat. no.:
    60141-124
  • Depth:
    4.8 cm (17/8")
  • Width:
    20.6 cm (81/8)"

Specifications

About this item

Cylinder wall-mount brackets are constructed of cast aluminum and contoured to allow cylinders to fit firmly along the support edge.

  • Cast aluminum construction
  • Nickel-plated, non-slip spring catch and buckle
  • Easily adjustable

Recessed screw holes at each side of the bracket allow for easy wall mounting. Features a 2.5 cm (1") wide, 137.2 cm (54") long nylon strap with nickel-plated, non-slip spring catch and buckle for fast, easy adjustment. For cylinders from 10.2 – 35.6 cm (4 – 14") in diameter. Available with or without a “Secure Cylinder” safety message strap.

Customers Who Bought This Also Bought