Order Entry
ContactUsLinkComponent
VWR* Talon* Rods, Aluminum
VWR® Talon® Rods, Aluminum
Catalog #: 60079-348
Supplier:  VWR International
Product available on GSA Advantage®
VWR* Talon* Rods, Aluminum
VWR® Talon® Rods, Aluminum
Catalog #: 60079-348
Supplier:  VWR International
Product available on GSA Advantage®

Specifications

  • Ø:
    13 mm (1/2")
  • Rod Length:
    152 mm (6")
  • Material:
    Aluminium
  • Cat. no.:
    60079-348

Specifications

About this item

These hard aluminum rods provide smooth fit with lab frames.

  • For use with VWR® Talon® lab frames
  • Resistant to corrosion
  • Precise fit
  • Chamfered ends

Center-less ground for precise fit. Corrosion-resistant with chamfered ends for easy location.

Customers Who Bought This Also Bought