Order Entry
ContactUsLinkComponent
KIMAX* Glass Stirring Rods
KIMAX® Glass Stirring Rods, Kimble Chase
Catalog #: 59060-047
KIMAX* Glass Stirring Rods
KIMAX® Glass Stirring Rods, Kimble Chase
Catalog #: 59060-047

Specifications

  • Ø:
    5 mm
  • Length:
    150 mm
  • Cat. no.:
    59060-047
  • Supplier No.:
    40500-150

Specifications

About this item

With rounded annealed ends

Designed from Federal Specification NNN-R-560, Type I requirements.

Customers Who Bought This Also Bought