Order Entry
ContactUsLinkComponent
KIMAX* Glass Stirring Rods
KIMAX® Glass Stirring Rods, Kimble Chase
Catalog #: 59060-025
KIMAX* Glass Stirring Rods
KIMAX® Glass Stirring Rods, Kimble Chase
Catalog #: 59060-025

Specifications

  • Ø:
    4 mm
  • Length:
    125 mm
  • Cat. no.:
    59060-025
  • Supplier No.:
    40500-125

Specifications

About this item

With rounded annealed ends

Designed from Federal Specification NNN-R-560, Type I requirements.

Customers Who Bought This Also Bought