VWR® Spinring® Magnetic Stir Bars
Catalog #: 58949-022
Supplier: VWR International
Specifications
- Coating:PTFE
- Ø:7.9 mm (5/16 ")
- Length:51 mm (2")
- Description:Spinring® 57.2 mm Ring O.D.
- Cat. no.:58949-022
Specifications
About this item
Octagonal stirring bar is contained within a removable stabilizing ring that centers it in a container and creates a deep vortex without splashing, chattering, or spinout.
- Removable stabilizing ring keeps bar centered
- Virtually eliminates spinout
- No chattering noise
- Autoclavable
Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.
Customers Who Bought This Also Bought