Order Entry
ContactUsLinkComponent
VWR* Spinbar* Stir Bars, Polygon
VWR® Spinbar® Magnetic Stir Bars, Polygon without Pivot Ring
Catalog #: 58948-985
Supplier:  VWR International
VWR* Spinbar* Stir Bars, Polygon
VWR® Spinbar® Magnetic Stir Bars, Polygon without Pivot Ring
Catalog #: 58948-985
Supplier:  VWR International

Specifications

  • Coating:
    PTFE
  • Ø:
    9.5 mm (3/8 ")
  • Length:
    22 mm (7/8")
  • Cat. no.:
    58948-985
  • Shape:
    Polygon

Specifications

About this item

Polygon-shaped Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.

  • Suitable for general stirring applications
  • Durable
  • Inert

Polygon-shaped stir bars come without a pivot ring.

Customers Who Bought This Also Bought