Order Entry
ContactUsLinkComponent
VWR* Spinbar* Stir Bars, Polygon
VWR® Spinbar® Magnetic Stir Bars, Polygon without Pivot Ring
Catalog #: 58948-984
Supplier:  VWR International
VWR* Spinbar* Stir Bars, Polygon
VWR® Spinbar® Magnetic Stir Bars, Polygon without Pivot Ring
Catalog #: 58948-984
Supplier:  VWR International

Specifications

  • Coating:
    PTFE
  • Ø:
    7.9 mm (5/16 ")
  • Length:
    15 mm (⅝”)
  • Cat. no.:
    58948-984
  • Shape:
    Polygon

Specifications

About this item

Polygon-shaped Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.

  • Suitable for general stirring applications
  • Durable
  • Inert

Polygon-shaped stir bars come without a pivot ring.

Customers Who Bought This Also Bought