VWR® Spinbar® Micro Stir Bars, Assortment
Catalog #: 58948-069
Supplier: VWR International
Specifications
- Coating:PTFE
- Description:Stir Bar Set
- Cat. no.:58948-069
Specifications
About this item
Think small with a Spinpak® of six metric, flea-sized, PTFE resin-coated magnetic stir bars.
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating
- Durable
- Inert
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclavable
These products are very small, efficient stirring bars.
Ordering information: Includes two of each of 7.9x1.5mm (5/16x1/16"), 7.9x2mm (5/16x1/8"), and 10x3mm (7/16x3/16") sizes.
Customers Who Bought This Also Bought