Order Entry
ContactUsLinkComponent
VWR* Circulus* Stir Bars, Color-Coded
VWR® Circulus™ Magnetic Stir Bars, Color-Coded
Catalog #: 58947-929
Supplier:  VWR International
VWR* Circulus* Stir Bars, Color-Coded
VWR® Circulus™ Magnetic Stir Bars, Color-Coded
Catalog #: 58947-929
Supplier:  VWR International

Specifications

  • Coating:
    PTFE
  • Ø:
    7.9 mm (5/16 ")
  • Length:
    36 mm (17/16")
  • Color:
    Yellow
  • Cat. no.:
    58947-929

Specifications

About this item

These magnetic stir bars perform well with all problem liquids, highly viscous mixtures, metal particles in fluids, and round (convex) bottom vessels.

  • Excellent self-centering characteristics
  • Fast mixing action with high turbulence
  • Greater stability
  • Less drifting at higher speeds

Available in two lengths, 36 and 54 mm (1 7/16 and 2 1/8"), each is designed as a 8 mm (0.315”) diameter bar with solid, color-coded, 20 mm (0.79") diameter, Teflon® resin end discs.

Customers Who Bought This Also Bought