Order Entry
ContactUsLinkComponent
VWR* Spinbar* Stir Bars, Octagon
VWR® Spinbar® Magnetic Stir Bars, Octagon
Catalog #: 58947-124
Supplier:  VWR International
VWR* Spinbar* Stir Bars, Octagon
VWR® Spinbar® Magnetic Stir Bars, Octagon
Catalog #: 58947-124
Supplier:  VWR International

Specifications

  • Coating:
    PTFE
  • Ø:
    9.5 mm (3/8 ")
  • Length:
    22 mm (7/8")
  • Cat. no.:
    58947-124
  • Shape:
    Octagon

Specifications

About this item

Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.

  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
  • Pivot ring aids in reducing friction and chattering (58948-091 does not have a pivot ring)
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification

Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars.

Ordering information: 6-Assortment includes six Octagon Spinbar® Magnetic Stir Bars, 2 each size; 5/16 x 1" (8 x 25.4mm), 5/16 x 1-1/2" (8 x 38.1mm); 5/16 x 2" (8mm x 50.8mm).
12-Assortment: Includes two of each 12.7x7.9mm (1/2x5/16"), 15.9x7.9mm (5/8x5/16"), 25.4x9.5mm (1x3/8"), 38.1x9.5mm (1-1/2x3/8"), 50.8x9.5mm (2x3/8"), and 63.5x9.5mm (2-1/2x3/8") sizes.
Customers Who Bought This Also Bought