Order Entry
ContactUsLinkComponent
VWR* Polypropylene Cuvette Rack
VWR® Cuvette Racks, PP
Catalog #: 58020-105
Supplier:  VWR International
VWR* Polypropylene Cuvette Rack
VWR® Cuvette Racks, PP
Catalog #: 58020-105
Supplier:  VWR International

Specifications

  • Description:
    Stackable Rack
  • Cat. no.:
    58020-105
  • L×W×H:
    11.4 x 11.1 x 11.1 cm (41/2 x 43/8 x 43/8")

Specifications

About this item

These two-tier racks support cuvettes with a minimum of physical contact, greatly reducing the chance of damage.

  • Racks accommodate 12 cuvettes of 10 mm light path
  • Racks will not scratch quartz cuvettes
  • Rack 58020-105 features Stack Rack™ Interlocking end plates for secure stacking
  • Manufactured from polypropylene, and steam Autoclavable at 121°C (250°F)

Only the corners of the cuvettes touch the racks, minimizing chance for contamination or damage.

Customers Who Bought This Also Bought