20X20 RPSF 500U 25/BOX BX25
Catalog #: 57457-104
Supplier: Analtech
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
$649.44 to $2,496.86
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Bard-Parker® Stainless Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Supplier: Aspen Surgical Products
$102.63 to $271.58
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Ammonium carbonate 30,0% (NH₃ basis), lumps, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$267.72 to $843.18
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36