ContactUsLinkComponent
Bonding/Grounding Wires
Uninsulated Bonding/Grounding Wires, New Pig
Catalog # 56620-208
Supplier:  New Pig
undefined
Uninsulated Bonding/Grounding Wires, New Pig
Catalog # 56620-208
Supplier:  New Pig
Supplier Number:  DRM495
Specifications
  • Description:
    Alligator clip grounding end, alligator clip bonding end, 91.4 cm (3')
  • Cat. no.:
    56620-208
  • Supplier No.:
    DRM495
Specifications
About this item
Reduces static sparks generated by the movement and flow of flammable liquids
  • Reduces static sparks generated by the movement and flow of flammable liquids
  • Helps you meet 29 CFR 1910.106 (e)(6)(ii) for grounding and bonding when transferring Class I flammables
Helps you meet 29 CFR 1910.106 (e)(6)(ii) for grounding and bonding when transferring Class I flammables.
Frequently Bought Together