SparkPlugs® Earplugs, Moldex®
Catalog #: 56610-677
Supplier: Moldex/Metric
Small Business Enterprise
Specifications
- NRR:31 dB
- Description:Earplugs
- Environmentally Preferable:Safer Human and Environmental Health
- Cat. no.:56610-677
- Supplier No.:6604
Specifications
About this item
Randomly multi-colored earplugs increase worker compliance
- Tapered shape ensures easy insertion and removal
- Shaped to fit even small ear canals
- Disposable
Extra soft, low pressure foam provides comfortable fit.
Customers Who Bought This Also Bought