SP Bel-Art Acid and Solvent Bottle Carriers
Catalog #: 56608-864
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacity:4 L (1 gal.)
- Ø:20 cm (8")
- Description:LDPE solvent carrier cone style
- Height:37 cm (14⅜")
- Cat. no.:56608-864
Specifications
About this item
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles.
- Cone-style lid has 16 mm (5/8") opening in the top for insertion of tubing, simplifies the transfer of liquids without pouring
- Truncated style lid permits pouring without the removal of the lid
- Designed to hold standard US reagent bottles
These acid and solvent bottle carriers are made of LDPE.
Customers Who Bought This Also Bought