B/T Sure™ Biological Indicators, Thermo Scientific
Catalog #: 55710-014
Supplier: Thermo Fisher Scientific
Specifications
- Description:B/T Sure™ Biological Indicator, Spore Population 105
- Cat. no.:55710-014
- Supplier No.:AY759X1
Specifications
About this item
Indicators verify the sterility of instruments, vials, glassware, and other products in less time than it takes to send samples to an outside lab.
- Use to verify sterility
- Purple color ensures sterilization
Purple color ensures sterilization. Color change from purple to yellow indicates an ineffective sterilization process. Indicators are supplied in a variety of packaging options, with each vial containing a spore population of either 105 or 106.
Customers Who Bought This Also Bought