Order Entry
ContactUsLinkComponent
DUOSEAL* Pump Oil
Accessories for DUOSEAL® Pumps
Catalog #: 54996-230
Supplier:  Welch Vacuum
Product available on GSA Advantage®
DUOSEAL* Pump Oil
Accessories for DUOSEAL® Pumps
Catalog #: 54996-230
Supplier:  Welch Vacuum
Product available on GSA Advantage®

Specifications

  • Description:
    Pump Oil, 3.78 L (1 gal.)
  • Cat. no.:
    54996-230
  • Supplier No.:
    1407K-15

Specifications

About this item

DUOSEAL® pump oil recommended for DUOSEAL® vacuum pumps

Exhaust filters trap exhaust oil mist and reduce pump noise. Supplied with housing and element. Replacement elements for filters also listed.

Features low vapor pressure, chemical stability, ideal viscosity, and no additives or inhibitors.

Customers Who Bought This Also Bought