SP Bel-Art Bacterial Cell Spreaders, Bel-Art Products, a part of SP
Catalog #: 53800-008
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Large
- Cat. no.:53800-008
- Blade Width:25.4 mm (1")
- Handle Length:20.3 cm (8")
- Supplier No.:377360009
Specifications
About this item
For spreading cells onto the surface of an agar plate to form individual colonies.
- Stainless steel construction
- Autoclavable
- May also be sterilized by flame
- Smooth triangular end
These stainless spreaders have a smooth triangular end with a straight spreading bar. The small size is ideal for 6 cm plates and the large size is ideal for 9 cm plates. Spreaders are autoclavable and may be flame sterilized.
Customers Who Bought This Also Bought