OBSERVATION TUBE 200 MM
Catalog #: 53673-021
Supplier: Koehler
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
214,10 $ to 1 039,32 $
Fluoride Standards, 1 ml = 1 mg F-, 1000 ppm F-, RICCA Chemical Company
Supplier: Ricca Chemical
166,86 $