Costar® Reagent Reservoirs, Polystyrene, Sterile, Corning
Catalog #: 53504-035
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Capacity:50 ml
- Sterility:Sterile
- Color:White
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:53504-035
- Supplier No.:4870
Specifications
About this item
White PS, sterile, disposable.
- Ideal for repetitively filling multi channel pipettors
- Manufactured from virgin PS
Customers Who Bought This Also Bought