Pure™ 1000 SoftFit™ Pipette Tips, Molecular BioProducts
Catalog #: 53225-708
Supplier: Thermo Scientific
Specifications
- Length:7.9 cm (31/8")
- Volume:100 - 1000 µL
- Cat. no.:53225-708
- Supplier No.:3580
Specifications
About this item
The SoftFit design of these tips ensures tip-to-tip precision and universal fit.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
- Nonsterile tips are autoclavable
This feature also makes loading and ejecting easy and minimizes hand fatigue. Tips are compatible with most major brand pipettors.
Ordering information: Tips 53225-712 and -710 are packaged 100 per tray, 8 trays per pack. Tips 53225-708 are bulk packaged.
Customers Who Bought This Also Bought