Pure™ 200G Pipette Tips, Molecular BioProducts
Catalog #: 53225-682
Supplier: Thermo Scientific
Specifications
- Length:4.97 cm (131/32")
- Volume:40 - 200 µl
- Sterile:+
- Supplier No.:3531
Specifications
About this item
The Pure 200G tips feature a large orifice designed for pipetting fragile cells, viscous samples, and genomic DNA.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
- Nonsterile tips are autoclavable
Tips have a volume range of 40 to 200 µl for larger samples. Compatible with Pipetman*, Nichiryo/Benchmate*, Eppendorf*, and others.
Ordering information: Tips 53225-682 and -680 are packaged 96 per tray, 10 trays per pack. Tips 53225-678 are bulk packaged.
Customers Who Bought This Also Bought