Pure™ Ultra-Micro Pipette Tips, Molecular BioProducts
Catalog #: 53225-671
Supplier: Thermo Scientific
Specifications
- Length:4.57 cm (113/16")
- Volume:0.5 - 20 µl
- Supplier No.:3520
Specifications
About this item
These tips are ideal for use with low volume samples.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
- Non-sterile tips are autoclavable
The Ultra-Micro style allows for increased accuracy and repeatablility at low volumes. The longer tip length easily reaches sample tube bottoms. For use with Eppendorf*, Nichiryo*, and Transferpette* pipettors.
Ordering information: Tips 53225-674 are packaged 96 per tray, 10 trays per pack. Tips 53225-671 are bulk packaged.
Customers Who Bought This Also Bought