MBP Flip-Strip™ Racks
Catalog #: 53225-604
Supplier: Thermo Scientific
Specifications
- Color:Fluorescent Orange
- Cat. no.:53225-604
- Supplier No.:8678
Specifications
About this item
This reversible rack is ideal for any researcher utilizing strip tubes for PCR.
- Made from recycled plastic
- Stable, low-profile design with an alphanumeric grid for easy tube identification
- FlipStrip racks come complete with a snug fitting lid
- Lid protects samples and allows racks to be stacked
- Several colors available
Customers Who Bought This Also Bought