PENETROMETER CONE HALF
Catalog #: 52934-607
Supplier: Koehler
Customers Who Bought This Also Bought
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Supplier: Ricca Chemical
205,73 $
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Polypropylene, Avantor®
Supplier: VWR International
45,66 $ to 47,08 $
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
147,27 $ to 464,44 $
Candida, Chromogenic medium, recommended for the isolation and identification of yeasts, allows for the differentiation of C. albicans, C. tropicalis and C. krusei based on differences in colony morphology and color, facilitates the detection of mixed yeast cultures, 15x100mm plate
Supplier: HARDY DIAGNOSTICS CA
67,41 $
Sodium phosphate dibasic, anhydrous ≥99.0%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
521,55 $