DISH HOLDER
Catalog #: 52933-557
Supplier: Koehler
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Poly-L-Lysine Coated and Silane Treated Microscope Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
177,13 $ to 177,13 $
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
156,54 $ to 571,97 $
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
178,29 $
Reagent Reservoirs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
277,83 $ to 277,83 $
Halt™ Protease and Phosphatase Inhibitor Cocktails, Thermo Scientific
Supplier: Thermo Scientific
2 074,46 $