Nalgene® Marking Pens, Thermo Scientific
Catalog #: 52887-008
Supplier: Thermo Fisher Scientific
Specifications
- Point Type:Fine
- Color:Black
- Description:Marking pens
- Cat. no.:52887-008
- Supplier No.:6310-0010
Specifications
About this item
Permanent black ink, pocket clip felt-tip pens for writing on glass and plastic surfaces
- Markings will not be affected by autoclaving
Customers Who Bought This Also Bought