Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Catalog #: 51280-114
Supplier: Thermo Fisher Scientific
Specifications
- Cat. no.:51280-114
- Dimensions:8.5×11"
- Supplier No.:6304-9811
Specifications
About this item
Nalgene® Polyolefin Plastic Paper Sheets roll or fold without cracking and stay flexible at cold temperatures
They won't rip, stretch or shrink and can be punched for use in binders or cut into labels. This paper is resistant to many solvents including acetone, formaldehyde, formalin, xylene, acetic acid and ethanol. They accept many inks including laser, inkjet, copier, markers, ballpoint, gels and technical markers. These sheets DO NOT accept pencil.
Customers Who Bought This Also Bought