Order Entry
ContactUsLinkComponent
VWR* Micro Cover Glasses, Round, No. 2
VWR® Micro Cover Glasses, Round, No. 2
Catalog #: 48382-063
Supplier:  VWR International
VWR* Micro Cover Glasses, Round, No. 2
VWR® Micro Cover Glasses, Round, No. 2
Catalog #: 48382-063
Supplier:  VWR International

Specifications

  • Ø:
    22 mm
  • Numbering:
    #2
  • Description:
    Cover glasses, round
  • Cat. no.:
    48382-063

Specifications

About this item

These microscope cover glasses are corrosion-resistant and uniformly thin and flat.

  • Borosilicate glass fabrication
  • Thickness: 0.17-0.25mm

The processing techniques result in superior cleanliness. Manufactured from borosilicate glass, cover glasses are free from bubbles, scratches, and striations.

Ordering information: Packaged 1 oz. per plastic box.
Customers Who Bought This Also Bought