SP Bel-Art Non sterile and Sterileware® Sampling Spatulas, Bel-Art Products, a part of SP
Catalog #: 47750-522
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacity:15 cm³
- Length:22.9 cm (9")
- Description:Nonsterile 9" Spatula
- Blade Length:152 mm (6")
- Handle Length:7.6 cm (3")
- Supplier No.:378400000
Specifications
About this item
These V-shaped spatulas are ideal for every type of powdered or granulated material.
- Available sterile or non sterile
- Molded from FDA-grade materials
- Safe for food, drug, and cosmetic contact
- V shape is ideal for powdered or granulated material
The sturdy point is perfect for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.
Long 14" spatula is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 18 mm (³/₄").
These one-piece spatulas are molded from FDA-grade materials and are safe for food, drug, and cosmetic contact.
Sterileware® Spatulas (9 and 14") are made of high-impact polystyrene, individually packaged, sterilized by gamma irradiation, lot stamped, and shipped with a Certificate of Processing. Non sterile 9" spatulas are made of polypropylene.
Customers Who Bought This Also Bought