ContactUsLinkComponent
Universal Fit Pipet Filtered Tip
Axygen® Hydrophobic Filter Barrier Pipette Tips, Corning
Catalog #: 47745-492
Supplier:  Corning
Product available on GSA Advantage®
Axygen® Hydrophobic Filter Barrier Pipette Tips, Corning
Catalog #: 47745-492
Supplier:  Corning
Product available on GSA Advantage®
Specifications
  • Volume:
    10 µl
  • Packed:
    96/Rack
  • Tip Style:
    Extended length
  • For Use With:
    Extended Length MicroVolume Tips For Use With Most Research-Grade Pipettors
  • Supplier No.:
    TXLF-10-R-S
Specifications
About this item
Filtered tips are ideal for molecular and microbiology applications in which carryover contamination can invalidate results.
Tips are designed to fit a wide variety of single-channel and multichannel pipettors. Flexible walls and a series of internal sealing rings ensure a secure fit and require less force to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
  • Tips are metal-free
  • Filters are made from polyethylene and contain no PCR inhibitors
Filtered tips are ideal for molecular and microbiology applications in which carryover contamination can invalidate results.
Tips are designed to fit a wide variety of single-channel and multichannel pipettors. Flexible walls and a series of internal sealing rings ensure a secure fit and require less force to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
Tips are designed to fit a wide variety of single-channel and multichannel pipettors. Flexible walls and a series of internal sealing rings ensure a secure fit and require less force to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
All tips, except Extended Length MicroVolume Tips For Use With Most Research-Grade Pipettors bulk pack, are presterilized by e-beam radiation for protection from contamination.
The filter position allows for sufficient space between the sample and filter at the maximum rated volume and in case of overpipetting, the sample can be easily recovered by dispensing.
Certifications: Certified RNase-, DNase-, and endotoxin-free.
Ordering information: To order non sterile tips, contact your sales representative.
Customers Who Bought This Also Bought