Order Entry
ContactUsLinkComponent
Assay Plate
Axygen® Deep Well and Assay Plates, Corning
Catalog #: 47743-984
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Assay Plate
Axygen® Deep Well and Assay Plates, Corning
Catalog #: 47743-984
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®

Specifications

  • Sterility:
    Sterile
  • Well Volume:
    550 µl
  • Color:
    Clear
  • Description:
    Round-Bottom Plates
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Packaging:
    50/case
  • Well Bottom Shape:
    Round
  • Supplier No.:
    P-96-450R-C-S

Specifications

About this item

Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.

  • 0.6 ml deep well plate
  • ANSI/SLAS 1-2004: Microplates - footprint dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film

Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.

Customers Who Bought This Also Bought