Accessories for Barnstead RO Systems, Thermo Scientific
Catalog #: 47742-616
Supplier: Thermo Fisher Scientific
Accessories for Barnstead RO Systems, Thermo Scientific
Catalog #: 47742-616
Supplier: Thermo Fisher Scientific
Specifications
- Description:MPS Cartridge
- Cat. no.:47742-616
- Supplier No.:D502114
Specifications
About this item
Customers Who Bought This Also Bought
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Supplier: Medicom
$49.54 to $495.19
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
$90.42 to $94.61
Methanol ≥99.9% (by GC, corrected for water content), BAKER ANALYZED™ HPLC for liquid chromatography, for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$491.55 to $1,692.92