VWR® Disposable Pasteur Pipettes, Glass, Plugged
Catalog #: 47735-748
Supplier: VWR International
Specifications
- Material:Borosilicate glass, 3.3 expansion
- Overall Length:14.6 cm (5¾")
Specifications
About this item
These disposable glass pipettes are furnished with cotton plugs.
- Disposable
- Glass fabrication
- Plugged
A positive constriction at the top holds plug in place.
Customers Who Bought This Also Bought