VWR® Caps for VWR® 9 mm Screw-Thread Vials
Catalog #: 46610-712
Supplier: VWR International
Specifications
- Cap Size:9 mm
- Cap Type:Polypropylene
- Septum:Red PTFE/White Silicone
- Description:Screw Caps
- Cap Color:Black
Specifications
About this item
Customers Who Bought This Also Bought
Potassium dihydrogen phosphate ≥99% (by acidimetry), GR ACS, Supelco®
Supplier: MilliporeSigma
$1,033.55 to $4,052.62
Masterflex® L/S® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Supplier: VWR International
$268.60
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$78.35 to $1,063.43
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
$150.16 to $1,502.77
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Sodium chloride ≥99.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$110.33 to $110.33
TruBond™ Adhesion Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$123.32 to $123.32
Potassium bromate ≥99.8% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$291.16