Nalgene® Dropper Bottles, Low-Density Polyethylene, Thermo Scientific
Catalog #: 46300-592
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:4 ml (0.14 oz.)
- Description:Natural Dropper Bottles with White Closures
- Cat. no.:46300-592
- Supplier No.:2750-9125
Specifications
About this item
Drop volume averages 40 µl per drop, depending on reagent used. Both natural and white dropper bottles have excellent chemical resistance and have been rated suitable for most biotech, diagnostic, and pharmaceutical applications. Natural, clear dispensing bottles permit easy content identification, while white dispensing bottles protect UV light-sensitive products. Closures are polypropylene and are available in white or assorted colors (five each of white, red, yellow, green, blue) for easy reagent identification.
Dropper bottles provide accurate, repeatable dispensing of various reagents and offer an alternative to pipetting.
Ordering information: Each pack includes tips, closures, and bottles ready for assembly.
Customers Who Bought This Also Bought