Automation Pipette Tips for Tecan® Workstations, Porex BioProducts
Catalog #: 46300-428
Supplier: Thermo Scientific
Specifications
- Volume:50 µl
- Tip Color:Natural
- Packed:96/Rack, 10 Racks
- Filter:+
- Sterile:+
- Tip Style:With Filter
- Supplier No.:LMF172-96RS-50
Specifications
About this item
Tips are compatible with, but not limited to, the Tecan® Genesis workstation.
Volumes range from 10 to 1000 µl. Available in two different colors, natural and black. Also available with or without filter. The flexible polypropylene design provides optimal fit for the workstation. Sterile tips are tray-packaged.
Customers Who Bought This Also Bought