VWR® CryoCooler Freeze Controller
Catalog # 414004-284
Supplier: VWR International
Specifications
- Description:VWR® CryoCooler Freeze Controller
- Cat. no.:414004-284
Specifications
About this item
When filled with isopropyl alcohol and placed in mechanical freezer, this polycarbonate freeze controller provides the -1°C per minute cooling rate required to successfully preserve cells.
- Non-mechanical
- Successfully preserves cells
- Insert helps prevent contamination
Floating rigid insert helps prevent contamination. Withstands repeated use. Holds up to eighteen 1.2–2 mL cryogenic vials.
Frequently Bought Together