Order Entry
ContactUsLinkComponent
VWR BOTTLE WASH 125ML PK6
VWR® Wash Bottles
Catalog #: 414004-225
Supplier:  VWR International
VWR BOTTLE WASH 125ML PK6
VWR® Wash Bottles
Catalog #: 414004-225
Supplier:  VWR International

Specifications

  • Cap Size:
    24 - 410 mm
  • Capacity:
    125 ml (4.2 oz.)
  • Cat. no.:
    414004-225

Specifications

About this item

LDPE wash bottle features a polypropylene screw cap and stem that are molded into a one-piece unit for leakproof operation.

  • LDPE construction
  • Provided with a polypropylene screw cap
  • Leakproof operation

Dispensing tube produces a fine stream, but can be cut for a coarser flow.

Customers Who Bought This Also Bought