Order Entry
ContactUsLinkComponent
VWR Plastic Bottles
VWR® Wide Mouth Bottles, Rectangular, HDPE,
Catalog #: 414004-193
Supplier:  VWR International
VWR Plastic Bottles
VWR® Wide Mouth Bottles, Rectangular, HDPE,
Catalog #: 414004-193
Supplier:  VWR International

Specifications

  • Cap Size:
    48-415
  • Capacity:
    500 ml (16.9 oz.)
  • Cat. no.:
    414004-193
  • W×L×H:
    64×99.5×114 mm

Specifications

About this item

Space-saving, leakproof bottles are for sampling, storage, or shipment of dry chemicals or liquids.

  • Sturdy construction
  • Space-saving design
  • Leakproof
  • HDPE fabrication

Sturdy construction prevents bulging, translucent material allows for optical clarity, and wide mouth makes filling easy. Bottles have excellent chemical resistance to most acids, bases, and alcohols, and can be stored in freezer to −100 °C (−148 °F). Bottle 414004-195 features integrated handle for better grip, greater safety, and pouring convenience. Polypropylene screw caps included.

Customers Who Bought This Also Bought