VWR® Large Wide Mouth Square Bottle, HDPE
Catalog #: 414004-190
Supplier: VWR International
Specifications
- Capacity:4 L (1 gal.)
- Dimensions:27.9H x 14D x 14W cm (11 x 51/2 x 51/2")
- Cat. no.:414004-190
Specifications
About this item
Leakproof, space-saving 3.8 L (1 gal.) bottle is ideal for collecting, storing, and transporting dry chemicals, environmental samples, reagents, and distilled water.
- Leakproof
- Space-saving design
- Wide mouth for easy access to contents
Can be used for low-temperature applications and stored in freezer to −100 °C (−148 °F). Features integrated handle for better grip, greater safety, and pouring convenience.
Ordering information: 100-415 white polypropylene cap included, assembled.
Customers Who Bought This Also Bought